missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FABP3 (H-FABP) (Muscle and Heart) Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75545
This item is not returnable.
View return policy
Description
Muscle-Cardiac FABP (human) Recombinant Protein, RP-75545, from Invitrogen.
This product contains purified recombinant human muscle and heart FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.Specifications
P05413 | |
PBS with 50% glycerol and no preservative | |
Human FABP3 | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Fatty Acid Binding Protein | |
FABP3 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control, Western Blot | |
2170 | |
10μg | |
N/A | |
RUO | |
Human FABP3 | |
Human | |
Hu | |
Recombinant | |
Recombinant |