missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor Xa Native Protein

Product Code. 11584141
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 11584141

Brand: Invitrogen™ RP43114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. RP-43114 contains native human Factor Xa protein. RP-43114, RP-43117, and RP43118 may be used as controls in many immunological assays. RP-43114 is the active version of Factor Xa. The EGR version (RP-43117) contains the tripeptide chloromethyl ketone inhibitor EGRck, which irreversibly blocks the active-site of Xa. The DEGR version (RP-43118) contains the fluorescent inhibitor Dansyl-EGRck (DEGR), which irreversibly blocks the active-site of Xa. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Factor Xa is a vitamin K-dependent glycoprotein that converts prothrombin to thrombin in the presence of factor Va, calcium and phospholipid during blood clotting.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00742
Concentration 1.8 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 2159
Molecular Weight (g/mol) 46 kDa
Name Human Factor Xa
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activated factor Xa heavy chain; Coagulation factor X; F10; F10a; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product
Common Name Factor Xa
Gene Symbol F10
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.