missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3A Antibody, R&D Systems™
Mouse Monoclonal Antibody
£146.86 - £333.68
Specifications
Antigen | FAM3A |
---|---|
Clone | 358112 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782477
|
R&D Systems
MAB2865-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18106041
|
R&D Systems
MAB2865 |
100 μg |
£333.68
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM3A Monoclonal specifically detects FAM3A in Human samples. It is validated for Western Blot.Specifications
FAM3A | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P98173 | |
60343 | |
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230 Accession # P98173 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FAM3A in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse FAM3A is observed and no cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, or rhFAM3D is observed. |
358112 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein, XAP-7 | |
FAM3A | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only