missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3A Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2865-SP
This item is not returnable.
View return policy
Description
FAM3A Polyclonal specifically detects FAM3A in Human samples. It is validated for Western Blot.Specifications
FAM3A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein, XAP-7 | |
Goat | |
Affinity Purified | |
RUO | |
60343 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
P98173 | |
FAM3A | |
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230 Accession # P98173 | |
25 μg | |
Primary | |
Detects human FAM3A in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, and rhFAM3D is observed. | |
Human | |
IgG |
For Research Use Only