missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FAM3A Antibody, R&D Systems™
Goat Polyclonal Antibody
£147.94 - £418.32
Specifications
Antigen | FAM3A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745688
|
R&D Systems
AF2865-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18073392
|
R&D Systems
AF2865 |
100 μg |
£418.32
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM3A Polyclonal specifically detects FAM3A in Human samples. It is validated for Western Blot.Specifications
FAM3A | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein, XAP-7 | |
FAM3A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P98173 | |
60343 | |
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230 Accession # P98173 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FAM3A in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, and rhFAM3D is observed. |
For Research Use Only