missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-16 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £2410.00
Specifications
Accession Number | O43320 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 8823 |
Molecular Weight (g/mol) | 23.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17830673
|
Gibco™
100-29-100UG |
100 μg |
£482.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830883
|
Gibco™
|
4 x 250 μg | N/A | ||||||
17861753
|
Gibco™
100-29-250UG |
250 μg |
£842.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17890943
|
Gibco™
100-29-25UG |
25 μg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870703
|
Gibco™
|
2 x 250 μg | N/A | ||||||
17820953
|
Gibco™
100-29-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.Specifications
O43320 | |
protein with no preservative | |
23.6 | |
E. coli | |
RUO | |
FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
8823 | |
Human FGF-16 | |
-20°C | |
<1 EU/μg | |
FGF16 | |
Unconjugated | |
AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |