missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-18 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-28-1MG
Additional Details : Weight : 0.16500kg
Description
FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures.Specifications
O76093 | |
protein with no preservative | |
20.1 | |
1 mg | |
-20°C | |
<1 EU/μg | |
Fgf18 | |
Unconjugated | |
AEENVDFRIH VENQTRARDD VSRKQLRLYQ LYSRTSGKHI QVLGRRISAR GEDGDKYAQL LVETDTFGSQ VRIKGKETEF YLCMNRKGKL VGKPDGTSKE CVFIEKVLEN NYTALMSAKY SGWYVGFTKK GRPRKGPKTR ENQQDVHFMK RYPKGQPELQ KPFKYTTVTK RSR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
8817 | |
Human FGF-18 | |
E. coli | |
RUO | |
D130055P09Rik; FGF; FGF18; FGF-18; Fibroblast growth factor; Fibroblast growth factor 18; UNQ420/PRO856; zFGF5 | |
Human | |
Recombinant | |
Lyophilized |