Learn More
Gibco™ Human FGF-21 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 100-42-100UG
 View more versions of this product
Product Code. 17880263
  • £482.00 / 100µg
Explore more special offers
Description
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9NSA1
protein with no preservative
19.5
100 μg
-20°C
<1 EU/μg
FGF21
Unconjugated
MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
≥ 90% by SDS-PAGE gel and HPLC analyses.
Functional Assay
26291
Human FGF-21
E. coli
RUO
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196
Human
Recombinant
Lyophilized