missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-acidic (FGF-1) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £1200.00
Specifica
Accession Number | P05230 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2246 |
Molecular Weight (g/mol) | 16.8 |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
17821533
|
Gibco™
100-17A-100UG |
100 μg |
£240.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17880113
|
Gibco™
100-17A-10UG |
10 μg |
£64.75
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800793
|
Gibco™
100-17A-1MG |
1 mg |
£1200.00
1mg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860953
|
Gibco™
100-17A-250UG |
250 μg |
£421.00
250µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820393
|
Gibco™
100-17A-500UG |
500 μg |
£720.00
500µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17801663
|
Gibco™
100-17A-50UG |
50 μg |
£157.00
50µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.Specifica
P05230 | |
protein with no preservative | |
16.8 | |
E. coli | |
RUO | |
acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
2246 | |
Human FGF-acidic (FGF-1) | |
-20°C | |
<1 EU/μg | |
FGF1 | |
Unconjugated | |
MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |