missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-basic (FGF-2/bFGF) (aa 1-155) Recombinant Protein

FGF-Basic (AA 1-155) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHG0264

 View more versions of this product

Product Code. 10008002

  • £89.25 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Human FGF-basic (FGF-2/bFGF) (aa 1-155) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

Fibroblast growth factor-basic is known as FGF-basic, bFGF, FGF 2, FGF-β, and heparin binding growth factor. FGF-basic is involved in a number of biological processes, including embryonic development and differentiation, neuron differentiation, survival, regeneration, and the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: proteins bioassayed with Gibco media
  • High-purity, recombinant, bioactive protein: to help ensure growth factors from Life Technologies are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/3T3 cells

Approved for shipment on Wet or Dry Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture
17.2 kDa
10 μg
Human
<0.1 ng/μg
E. coli
Recombinant, Ligand
FGFs (Fibroblast Growth Factors)
Gibco™
2247
Sequential Chromatography
FGF-Basic (AA 1-155)
Store in refrigerator (2–8°C).
Approved for shipment on Wet or Dry Ice
Growth Factors & Receptors
Lyophilized
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis
95%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.