Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF) Recombinant Protein
Human FGF-basic (FGF-2/bFGF) Recombinant Protein
Brand: Gibco™ 13256029
Code : 50
Additional Details : Weight : 1.00000kg
Description
Human FGF-basic (FGF-2/bFGF) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
bFGF (also known as fibroblast growth factor-basic, FGF-basic, FGF-β, FGF2, or heparin binding growth factor) is used in studies of angiogenesis, mitogenesis of fibroblasts, neurite outgrowth in PC-12 cells, receptor binding, and tyrosine protein kinase phosphorylation.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: proteins bioassayed with Gibco media
- High-purity, recombinant, bioactive protein: to help ensure growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; recombinant human bFGF specific activity is determined by thymidine incorporation into serum-starved Balb/c 3T3 cells
- Proven compatibility: Recombinant proteins are bioassayed with Gibcomedia
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
10 μg | |
Human | |
Carrier-Free | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis |
PAGE | |
FGF-Basic | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
E. coli | |
Full Length, Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.