Learn More
Applied Biosystems™ Human, Forward Primer, HPLC
Human, Forward Primer, HPLC
Brand: Applied Biosystems™
Additional Details : Weight : 0.00100kg
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Specifications
Human Forward HPLC Primer, 50 nmole, Solid, For PCR and Sanger Sequencing | |
Forward | |
HPLC | |
Solid |
Human Forward HPLC Primer | |
50 nmol | |
For PCR and Sanger Sequencing |
For Research Use Only. Not for use in diagnostic procedures.