missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Galectin-3 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £1200.00
Spezifikation
Accession Number | P17931 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3958 |
Molecular Weight (g/mol) | 26 |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
17834633
|
Gibco™
450-38-100UG |
100 μg |
£240.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863923
|
Gibco™
450-38-10UG |
10 μg |
£64.75
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17854233
|
Gibco™
450-38-1MG |
1 mg |
£1200.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17814623
|
Gibco™
450-38-250UG |
250 μg |
£421.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17884683
|
Gibco™
450-38-500UG |
500 μg |
£720.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17864003
|
Gibco™
450-38-50UG |
50 μg |
£157.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.Spezifikation
P17931 | |
protein with no preservative | |
26 | |
E. coli | |
RUO | |
35 kDa lectin; advanced glycation end-product receptor 3; AGE-R3; beta-galactosides-binding lectin; carbohydrate-binding protein 35; CBP 35; CBP30; CBP-30; CBP35; epsilon BP; gal3; gal-3; galactose-specific lectin 3; Galactoside-binding protein; GALBP; galectin 3; Galectin3; galectin-3; GALIG; GBP; IgE binding protein; igE-binding protein; L-29; L31; L-31; L-34; L-34 galactoside-binding lectin; Laminin-binding protein; lectin galactoside-binding soluble 3; lectin galactoside-binding soluble 3 protein; lectin L-29; lectin, galactose binding, soluble 3; lectin, galactoside binding soluble 3; lectin, galactoside-binding, soluble, 3; lectin, galactoside-binding, soluble, 3 (galectin 3); LGALS2; Lgals3; MAC2; Mac-2; Mac-2 antigen | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
3958 | |
Human Galectin-3 | |
-20°C | |
<1 EU/μg | |
LGALS3 | |
Unconjugated | |
ADNFSLHDAL SGSGNPNPQG WPGAWGNQPA GAGGYPGASY PGAYPGQAPP GAYPGQAPPG AYHGAPGAYP GAPAPGVYPG PPSGPGAYPS SGQPSAPGAY PATGPYGAPA GPLIVPYNLP LPGGVVPRML ITILGTVKPN ANRIALDFQR GNDVAFHFNP RFNENNRRVI VCNTKLDNNW GREERQSVFP FESGKPFKIQ VLVEPDHFKV AVNDAHLLQY NHRVKKLNEI SKLGISGDID LTSASYTMI | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |