missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gamma-Synuclein Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5745R-500
This item is not returnable.
View return policy
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
gamma-Synuclein | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6623 | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
514304R | |
Western Blot 0.5 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL | |
Q6FHG5 | |
SNCG | |
E. coli-derived recombinant human gamma-Synuclein Asn47-Glu120 Accession No. Q6FHG5 | |
500 ug | |
Primary | |
Detects human gamma-Synuclein in direct ELISAs and Western blots. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction