Learn More
R&D Systems™ Human GDF-11/BMP-11 DuoSet ELISA
Human GDF-11/BMP-11 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DY1958-05
Additional Details : Weight : 0.00970kg
Product Code. 30020801
  • £377.96 / 1 set
Estimated Shipment: 29-07-2024
to see stock.
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Description
Description
The Human GDF-11/BMP-11 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human GDF-11/BMP-11.
Specifications
Specifications
1 Kit (for 5 Plates)
Biotin
ELISA
BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11
Human
GDF-11/BMP-11
10220
Store the unopened product at 2°C to 8°C. Do not use past expiration date.