missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDNF Recombinant Protein
Glial-Derived Neurotrophic Factor (GDNF) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC7045
This item is not returnable.
View return policy
Description
Human GDNF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
GDNF is mainly known for its affect on neurons, but it is also involved in other biological processes including kidney development.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by inducing neurite outgrowth on PC-12 cells in the presence of recombinant soluble GFR-α
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment on Wet or Dry Ice.
Specifications
Cell Culture | |
30.1 kDa | |
10 μg | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Neurotrophic Factors | |
Gibco™ |
2668 | |
Sequential Chromatography | |
G-DNF | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation | |
98 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Spot an opportunity for improvement?Share a Content Correction