missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human GDNF Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Brand:  Gibco™ 450-10-500UG

Additional Details : Weight : 0.16500kg

 View more versions of this product

Product Code. 17874643

  • £2495.00 / 500µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
Specifications

Specifications

P39905
protein with no preservative
30.4
500 μg
-20°C
<1 EU/μg
GDNF
Unconjugated
MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
≥ 98% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay, Western Blot
2668
Human GDNF
E. coli
RUO
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor
Human
Recombinant
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers