missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1439
This item is not returnable.
View return policy
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Human samples. It is validated for Western Blot.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64096 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
215725 | |
Western Blot 1 ug/mL | |
NP_071422 | |
GFRA4 | |
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-4/GDNF R alpha-4 Asn24-Ser245 (Ser140Cys)(predicted) Accession # NP_071422 | |
500 μg | |
Primary | |
Detects human GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) GFR alpha-1, recombinant rat GFR alpha-1, rhGFR alpha-2, rhGFR alpha-3, or recombinant mouse GFR alpha-4 is observed. | |
Human | |
Purified |
For Research Use Only