missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GLP-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 130-08-5MG
This item is not returnable.
View return policy
Description
GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.Specifications
P01275 | |
protein with no preservative | |
3.3 | |
5 mg | |
-20°C | |
<1 EU/μg | |
GCG | |
Unconjugated | |
HAEGTFTSDV SSYLEGQAAK EFIAWLVKGR G | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
2641 | |
Human GLP-1 | |
E. coli | |
RUO | |
GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction