missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GM-CSF Quantikine ELISA Kit
Human GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
£534.54 - £3066.87
Specifications
Assay Range | 7.80 - 500pg/mL |
---|---|
Assay Sensitivity | 3pg/mL |
Conjugate | HRP |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Specificity | Natural and Recombinant Human GM-CSF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731944
|
R&D Systems™
DGM00 |
1 Plate |
£534.54
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771904
|
R&D Systems™
SGM00 |
6 Plates |
£3066.87
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Human GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human GM-CSF in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).Specifications
7.80 - 500pg/mL | |
HRP | |
Natural and Recombinant Human GM-CSF | |
ELISA | |
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL) | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim |
3pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
Human | |
1437 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |