missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GM-CSF Recombinant Protein
Recombinant Human Granulocyte Macrophage Colony-Stimulating Factor (GM-CSF) is a bioactive protein intended for use in cell culture applications
£192.00 - £2790.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 1437 |
Molecular Weight (g/mol) | 14 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | GM-CSF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10797823
|
Gibco™
PHC2015 |
10 μg |
£192.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10150813
|
Gibco™
PHC2011 |
100 μg |
£1538.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10552045
|
Gibco™
PHC2013 |
1 mg |
£2790.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human GM-CSF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Specifications
Cell Culture | |
14 kDa | |
GM-CSF | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
GM-CSF, G-CSF, & M-CSF | |
Gibco™ |
1437 | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title