Learn More
Invitrogen™ Human GM130 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP296
Product Code. 10839230
  • £154.00 / Each
Explore more special offers
Description
Description
GM130/GOLGA2 Synthetic Peptide, PEP-296, from Invitrogen™.
GM130 is a putative cis-Golgi matrix protein that belongs to the golgin family of autoantigens possessing extensive coiled-coil domains. This protein interacts with GRASP65 and acts as a receptor for the vesicle docking protein p115 during vesicle fusion. This peptide corresponds to amino acids G(368) QVMESVRQLQMERDK (383) of human GM130 and can be used as a blocking peptide with polyclonal antibody PA1-077.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2801
purified
Human GM130
-20°C
Lyophilized
Blocking Assay, Control
GOLGA2
No preservative
50 μg
Human
Q08379
1 mg/mL
Synthetic Peptide
water with no preservative
Golgin subfamily A member 2, 130 kDa cis-Golgi matrix protein, GM130, GM130 autoantigen, Golgin-95, GOLGA2
Yes
Peptide
RUO - research use only