missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GNMT ELISA Kit

ELISA

Brand:  Invitrogen™ EH226RB

 View more versions of this product

Product Code. 17154513

  • £570.00 / Pack of 96
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Human GNMT quantitates human GNMT in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GNMT.

The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.1.1.20; epididymis secretory sperm binding protein Li 182 mP; folate-binding protein; glycine methyltransferase; glycine N methyl transferase; glycine N-methyltransferase; GNMT; HEL-S-182 mP
2.5 ng/mL
ELISA
Colorimetric Microplate Reader
GNMT
<10
HRP
RUO
2°C to 8°C
1 hrs 20 mins
2.5 to 500 ng/mL
HRP
Plasma, Serum, Supernatant
27232
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
GNMT
4 hrs 45 mins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.