missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GPIHBP1 (AAH35810) VersaClone cDNA
R&D Systems™ Human GPIHBP1 (AAH35810) VersaClone cDNA is a glycosylphosphatidylinositolanchored protein that is a member of the lymphocyte antigen 6 (Ly6) family.
Brand: R&D Systems™ RDC1987
Additional Details : Weight : 0.00970kg
Description
The Human GPIHBP1 (AAH35810) VersaClone cDNA clone contains the complete ORF for GPIHBP1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPIHBP1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
hGPIHBP1 | |
10 μg at 0.2 μg/μL | |
568bp | |
338328 | |
Human |
AAH35810 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |