missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPVI Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF3627-SP
This item is not returnable.
View return policy
Description
GPVI Polyclonal specifically detects GPVI in Human samples. It is validated for Western Blot.Specifications
GPVI | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI | |
Sheep | |
Affinity Purified | |
RUO | |
51206 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q9HCN6 | |
GP6 | |
Mouse myeloma cell line NS0-derived recombinant human GPVI (R&D Systems, Catalog # 3627-GP) Gln21-Lys267 Accession # Q9HCN6 | |
25 μg | |
Primary | |
Detects human GPVI in direct ELISAs and Western blots. | |
Human | |
IgG |
For Research Use Only