missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPVI Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
£147.94 - £469.23
Specifications
Antigen | GPVI |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715157
|
R&D Systems
AF3627-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18012422
|
R&D Systems
AF3627 |
100 μg |
£469.23
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPVI Polyclonal specifically detects GPVI in Human samples. It is validated for Western Blot.Specifications
GPVI | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI | |
GP6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9HCN6 | |
51206 | |
Mouse myeloma cell line NS0-derived recombinant human GPVI (R&D Systems, Catalog # 3627-GP) Gln21-Lys267 Accession # Q9HCN6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human GPVI in direct ELISAs and Western blots. |
For Research Use Only