missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human HCC-1 (CCL14) (66 aa) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-38B-500UG
This item is not returnable.
View return policy
Description
This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.Specifications
Q16627 | |
protein with no preservative | |
7.8 | |
500 μg | |
-20°C | |
<1 EU/μg | |
CCL14 | |
Unconjugated | |
GPYHPSECCF TYTTYKIPRQ RIMDYYETNS QCSKPGIVFI TKRGHSVCTN PSDKWVQDYI KDMKEN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
6358 | |
Human HCC-1 (CCL14) (66 aa) | |
E. coli | |
RUO | |
C-C motif chemokine 14; C-C motif chemokine ligand 14; CC-1; CC-3; CCL14; chemokine (C-C motif) ligand 14; chemokine CC-1/CC-3; chemokine CC-3; CKB1; H HCC-1(1-74); HCC-1; HCC-1(1-74); HCC-1(3-74); HCC-1(4-74); HCC-1(9-74); HCC-1/HCC-3; HCC-3; hemofiltrate CC chemokine 1; MCIF; NCC2; NCC-2; new CC chemokine 2; SCYA14; SCYL2; small inducible cytokine subfamily A (Cys-Cys), member 14; small-inducible cytokine A14; SY14 | |
Human | |
Recombinant | |
Lyophilized |