Learn More
Invitrogen™ Human HDAC3 (aa 415-428) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP071
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human HDAC3 (residues 415-428) Synthetic Peptide, PEP-071, from Invitrogen.
PEP-071 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 415-428 of human HDAC 3 with an amino terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C-D415-G-D-H-D-N-D-K-E-S-P-V-E-I428This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human HDAC 3, catalog &Num; PA1-862. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 50μL of distilled water for a concentration of 1mg/mL.Specifications
8841 | |
purified | |
Human HDAC3 (aa 415-428) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
O15379 | |
1 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
HDAC3 | |
No preservative | |
50 μg | |
C - D415 - G - D - H - D - N - D - K - E - S - P - V - E - I428 |