missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Heat Stable bFGF Recombinant Protein

Product Code. p-8632000
Click to view available options
Quantity:
100 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 16443569

Brand: Gibco™ PHG0360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to a concentration of 1.0 mg/mL. Alternatively, reconstitute at lower concentrations in carrier-containing medium. Do not vortex. Apportion the reconstituted protein into working aliquots and store at -20°C. Make any further dilutions of the reconstituted protein in medium, or buffered solution containing carrier protein, such as PBS with 0.1% BSA. For the 5 μg unit size, reconstitute directly in carrier-containing medium. For other unit sizes, reconstitute to 1.0 mg/mL as follows: 50 μg, reconstitute in 50 μL 100 μg, reconstitute in 100 μL 500 μg, reconstitute in 500 μL Storage: Store the lyophilized protein at -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at -20°C (not in a frost-free freezer) for up to 12 months. Avoid repeated freeze-thaw cycles.

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09038
For Use With (Application) Bioactivity
Formulation 20mM potassium phosphate with 750mM NaCl and no preservative
Gene ID (Entrez) 2247
Molecular Weight (g/mol) 19 kDa
Name Human Heat Stable bFGF
Purification Method Purified
Quantity 500 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); Fibroblast growth factor basic; HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin
Common Name FGF2
Gene Symbol FGF2
Protein Length 155 aa + 20 aa for N-terminal tag for purification purposes
Biological Activity ED50 = 0.6 - 1.1 ng/mL; determined by the dose-dependent proliferation of mouse NIH/3T3 fibroblast cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sterility Sterile
Sequence 155 aa + 20 aa for N-terminal tag for purification purposes
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 0.6 - 1.1 ng/mL; determined by the dose-dependent proliferation of mouse NIH/3T3 fibroblast cells.
Endotoxin Level <1 EU/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Growth Factors and Receptors
Form Lyophilized
Research Category Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.