Learn More
Invitrogen™ eBioscience™ Human HSP27 Platinum ELISA Kit
For the quantitative detection of human HSP27.
Brand: Invitrogen™ BMS2086TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes:
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP27
- Biotin-Conjugate anti-human HSP27 monoclonal antibody
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
10.9pg/mL | |
Human | |
10 x 96 Tests | |
hsp27 |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
Heat shock protein 27, heat shock protein beta-1, germ cell-dependent phosphoprotein-1 |