missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human HSP70/HSPA1A DuoSet ELISA
Human HSP70/HSPA1A DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY1663-05
Includes: Human HSP70 Capture Antibody; Human HSP70 Detection Antibody; Human HSP70 Standard; Streptavidin-HRP
Description
The Human HSP70/HSPA1A DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human HSP70/HSPA1A.Specifications
5 Plates | |
Biotin | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Human | |
ELISA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction