missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IFN-gamma Recombinant Protein
Recombinant Human Interferon-γ (IFN-γ) is a bioactive protein intended for use in cell culture applications
£194.00 - £2010.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 3458 |
Molecular Weight (g/mol) | 16.9 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IFN-γ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10474733
|
Gibco™
PHC4031 |
100μg |
£194.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10425143
|
Gibco™
PHC4033 |
1mg |
£2010.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human IFN-gamma Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone resorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytostasis of human WiDr cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
16.9 kDa | |
IFN-γ | |
Store in ultra-cold freezer (-68 to -85°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Interferons | |
Gibco™ |
3458 | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Dry Ice | |
Cytokines & Receptors | |
Liquid | |
Immunology, Signal Transduction, Stem Cell Research, Virology, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.