Learn More
STEMCELL Technologies Human IgE ELISA Antibody Pair Kit
For detection and measurement of human immunoglobulin E
Brand: STEMCELL Technologies 01993
This item is not returnable.
View return policy
Description
- The Human Immunoglobulin E (IgE) ELISA Antibody Pair Kit is intended for those who want the flexibility of setting up their own ELISA assay
- This kit includes capture and detection antibodies and an IgE standard
- It is designed for the quantitative detection and measurement of human IgE in biological fluids such as serum, plasma, and cell culture supernatants
- IgE is predominantly found in the lungs, skin, and mucous membranes and is the least common immunoglobulin in serum
- IgE plays a fundamental role in allergic reactions by stimulating degranulation of eosinophils, mast cells, and basophils through interaction with the Fc epsilon Receptor I (FcεRI)
- NOTE: This kit includes sufficient reagents for 6 x 96-well ELISA plates
- ELISA plates (Catalog #38019) and pNPP Substrate (Catalog #01917) are required for use with the Human IgE ELISA Antibody Pair Kit and are available for purchase separately
- For serum or plasma samples, ELISA Diluent (Catalog #01916) is also required
Specifications
B Cells; Hybridomas | |
1 Kit for 6 Plates |