missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IGF-BP1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 350-10-1MG
This item is not returnable.
View return policy
Description
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
P08833 | |
protein with no preservative | |
25.4 | |
1 mg | |
-20°C | |
<1 EU/μg | |
IGFBP1 | |
Unconjugated | |
MAPWQCAPCS AEKLALCPPV SASCSEVTRS AGCGCCPMCA LPLGAACGVA TARCARGLSC RALPGEQQPL HALTRGQGAC VQESDASAPH AAEAGSPESP ESTEITEEEL LDNFHLMAPS EEDHSILWDA ISTYDGSKAL HVTNIKKWKE PCRIELYRVV ESLAKAQETS GEEISKFYLP NCNKNGFYHS RQCETSMDGE AGLCWCVYPW NGKRIPGSPE IRGDPNCQIY FNVQN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
3484 | |
Human IGF-BP1 | |
E. coli | |
RUO | |
AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; hIGFBP-1; IBP 1; IBP1; IBP-1; IGF BP25; IGFBA; IGF-binding protein 1; IGFBP; IGFBP 1; Igfbp1; Igfbp-1; IGF-BP25; Insulin like growth factor binding protein; insulin like growth factor binding protein 1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12 | |
Human | |
Recombinant | |
Lyophilized |