missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IGF-I Recombinant Protein

Recombinant Human Insulin-like Growth Factor 1 (IGF1) is a bioactive protein intended for use in cell culture applications

£137.00 - £280.00

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 7.5 kDa
Gene Alias Igf-1
Species Human
Content And Storage Store in freezer (-5 to -30°C).
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12063599
View Documents
Gibco™
PHG0078
20 μg
£137.00
20µg
Please to purchase this item. Need a web account? Register with us today!
11682069
View Documents
Gibco™
PHG0071
100 μg
£280.00
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human IGF-I Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IGF1 binds its receptor, IGF1R, activating several signaling pathways. IGF1 has been shown to be involved in proliferation, differentiation, survival, growth, apoptosis and regeneration.
  • High purity: Minimizes interference from other proteins or contaminants
  • High biological activity: More results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: To help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High biological activity: In-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by its ability to stimulate the proliferation of mouse FDC-P1 cells
Specifications

Specifications

Cell Culture
Igf-1
Store in freezer (-5 to -30°C).
2 ng/ml
< 1 EU/ml
Growth Factors & Receptors
Lyophilized
Neurobiology, Drug Development, Oncology, Signal Transduction
7.5 kDa
Human
Carrier-Free
proliferation of mouse FDC-P1 cells
E. coli
Recombinant, Ligand
IGFs (Insulin-Like Growth Factors)
98 %
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.