missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IGF-II, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£79.50 - £1324.00
Specifications
Accession Number | P01344 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3481 |
Molecular Weight (g/mol) | 7.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17833413
|
Gibco™
AF-100-12-100UG |
100 μg |
£271.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17833313
|
Gibco™
AF-100-12-10UG |
10 μg |
£79.50
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863783
|
Gibco™
AF-100-12-1MG |
1 mg |
£1324.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17823713
|
Gibco™
AF-100-12-500UG |
500 μg |
£796.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17823313
|
Gibco™
AF-100-12-50UG |
50 μg |
£178.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.Specifications
P01344 | |
protein with no preservative | |
7.5 | |
E. coli | |
RUO | |
AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
3481 | |
Human IGF-II, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
IGF2 | |
Unconjugated | |
AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |