missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG Isotype Control, Agarose, Novus Biologicals™
Human Antibody
Brand: Bio-Techne NBP1-96830
Additional Details : Weight : 0.00970kg
Description
IgG specifically detects IgG in Human samples. It is validated for Immunoprecipitation, Immunoaffinity Purification.Specifications
IgG | |
Agarose | |
Immunoglobulin G, ImmunoglobulinG | |
Human | |
10 mg | |
Isotype Control (Primary) | |
Human | |
IgG |
2.5 mg/ml | |
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only