missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgG (Total) Human ELISA Kit

For the quantitative detection of human IgG.

Brand:  Invitrogen™ BMS2091TEN

2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 15591617

  • £3070.00 / Pack of 10
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human total IgG
This item is not returnable. View return policy

Description

Description

The Human Total Immunoglobulin G (Hu IgG) ELISA quantitates Hu IgG in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG. Principle of the method The Human IgG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody’s species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.6-100 ng/mL
HRP
Plasma, Serum
ELISA
2.6%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
10 x 96 Tests
Plasma, 10 μL; Serum, 10 μL
Immunoglobulin G
1 hr. 20 min.
0.24 ng/mL
ELISA Kit
Human
Colorimetric Microplate Reader
5.3%
HRP
RUO
2°C to 8°C
Human
1 hr. 30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only