Learn More
Invitrogen™ IgG3 Human ELISA Kit
For the quantitative detection of human IgG3.
£451.00 - £3070.00
Specifications
Conjugate | HRP |
---|---|
Sample Type | Plasma, Serum |
Target Species | Human |
For Use With (Application) | ELISA |
Synonym | Immunoglobulin G2 |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgG3
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15581627
|
Invitrogen™
BMS2094 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
15511637
|
Invitrogen™
BMS2094TWO |
2 x 96 Tests |
£618.00
Pack of 2 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15591627
|
Invitrogen™
BMS2094FIVE |
5 x 96 Tests |
£1548.00
Pack of 5 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15501637
|
Invitrogen™
BMS2094TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Description
The Human Immunoglobulin G3 (Hu IgG3) ELISA quantitates Hu IgG3 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG3. Principle of the method The Human IgG3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
HRP | |
Human | |
Immunoglobulin G2 |
Plasma, Serum | |
ELISA |
For Research Use Only