missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-1 alpha ELISA Kits

Measure levels of human interleukin-1 alpha (IL-1a) in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ EH2IL1A

 View more versions of this product

Product Code. 10066023

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Assay IL-1a (human interleukin-1 alpha) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-1 alpha ELISA Kits.

These human IL-1 alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-1 alpha (IL-1α) in samples at 50 per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-1 alpha (IL-1α)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 10.2 to 400pg/mL

  • Sensitivity: < 2pg/mL

  • Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IFNα, IFNγ, IL-1β, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, or mouse IL-1α. Human IL-1ra does not interfere with this ELISA

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10.2 to 400pg/mL
Human IL-1 alpha ELISA Kit
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
96 microplate wells
Absorbance at 450nm minus absorbance at 550nm
1 Plate Kit
Natural and recombinant human interleukin-1 alpha (IL-1a)
<2pg/mL
Colorimetric ELISA (TMB substrate) with Plates
No significant cross-reaction with human IFNa, IFNg, IL-1ß, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, or mouse IL-1a. Human IL-1ra does not interfere with this ELISA
Stop Solution, 13mL, Streptavidin-HRP Concentrate, 75µL, Standard Diluent, 12mL, contains 0.01% thimerosal, Biotinylated Antibody Reagent, 8mL, TMB Substrate Solution, 13mL, Anti-Human IL-1 alpha Precoated 96-well Strip Plate, 1 ea, HRP Dilution Buffer, 14mL, Lyophilized Recombinant Human IL-1 alpha Standard, 2 vials, Adhesive Plate Covers, 6 ea, 30X Wash Buffer, 50mL
Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
50 μL
3 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers