Learn More
R&D Systems™ Human IL-10 DuoSet ELISA
Human IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marque: R&D Systems™ DY217B-05
220.57 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Informations supplémentaires : Weight : 0.09700kg
Description
The Human IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-10.Spécification
ELISA Development Kit | |
Human | |
31.20 to 2000 pg/mL | |
ELISA | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
3586 | |
5 Plates |