missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-12 p70 Human Uncoated ELISA Kit with Plates

Specifically measures the bioactive, heterodimeric form of IL-12, p70, without interference by p40 monomer, homodimer, or IL-23 (p19/p40). Engineered for accurate and precise measurement of human IL-12 p70 protein levels from samples including serum, plasma, and cell culture supernatants.

£165.00 - £546.00

Specifications

Accession Number P29459,P29460
Assay Range 4-500 pg/mL
Assay Sensitivity 4 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Alert:

Contains Azide


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15591057
View Documents Promotion Details
Invitrogen™
88-7126-22
2 x 96 Tests
£165.00
Pack of 2
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
 
15501067
View Documents Promotion Details
Invitrogen™
88-7126-76
20 x 96 Tests
£546.00
Pack of 20
In Stock
to see stock.
Add to basket
 
15521067
View Documents Promotion Details
Invitrogen™
88-7126-86
10 x 96 Tests
£392.00
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

The Human Interleukin 12 p70 (IL-12 p70) Uncoated ELISA Kit contains pre-matched antibody pairs,plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-12 p70. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Human IL-12 p70 ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)
Specifications

Specifications

P29459,P29460
4 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
AA690181,IL-15RA,AI503618,IL-15
HRP
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL
Interleukin-12, IL12, IL23
1 hr. 45 min.
4-500 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
3592, 3593
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 µM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Streptavidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
RUO
2°C to 8°C
Human
24 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only