missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-13 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £4150.00
Specifications
Accession Number | P35225 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3596 |
Molecular Weight (g/mol) | 12.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17880593
|
Gibco™
200-13-100UG |
100 μg |
£832.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17801693
|
Gibco™
200-13-10UG |
10 μg |
£157.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830813
|
Gibco™
200-13-1MG |
1 mg |
£4150.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860593
|
Gibco™
200-13-250UG |
250 μg |
£1456.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17840893
|
Gibco™
200-13-2UG |
2 μg |
£64.75
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17811793
|
Gibco™
200-13-500UG |
500 μg |
£2495.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17841693
|
Gibco™
200-13-50UG |
50 μg |
£502.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
P35225 | |
protein with no preservative | |
12.6 | |
E. coli | |
RUO | |
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
3596 | |
Human IL-13 | |
-20°C | |
<1 EU/μg | |
Il13 | |
Unconjugated | |
MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGRFN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |