missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17A Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 200-17-50UG
This item is not returnable.
View return policy
Description
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
Q16552 | |
protein with no preservative | |
31.3 | |
50 μg | |
-20°C | |
<1 EU/μg | |
IL17A | |
Unconjugated | |
MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
3605 | |
Human IL-17 A | |
E. coli | |
RUO | |
Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17 A; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17 A; interleukin-17; Interleukin17A; interleukin-17 A; R-IL-17-A | |
Human | |
Recombinant | |
Lyophilized |