missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-17F Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £2400.00
Specifications
Accession Number | Q96PD4 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 112744 |
Molecular Weight (g/mol) | 30.1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17860733
|
Gibco™
200-25-100UG |
100 μg |
£480.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850913
|
Gibco™
200-25-1MG |
1 mg |
£2400.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830233
|
Gibco™
200-25-250UG |
250 μg |
£838.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17891873
|
Gibco™
200-25-25UG |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17801113
|
Gibco™
200-25-500UG |
500 μg |
£1436.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860433
|
Gibco™
200-25-50UG |
50 μg |
£291.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17851703
|
Gibco™
200-25-5UG |
5 μg |
£64.75
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.Specifications
Q96PD4 | |
protein with no preservative | |
30.1 | |
E. coli | |
RUO | |
C87042; CANDF6; cytokine ML-1; IL17F; IL-17 F; IL-24; ILN; Interleukin; interleukin 17 F; Interleukin17F; interleukin-17 F; interleukin-24; ML1; ML-1; mutant IL-17 F | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
112744 | |
Human IL-17 F | |
-20°C | |
<1 EU/μg | |
IL17F | |
Unconjugated | |
MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |