missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-18 Human ELISA Kit

For the quantitative detection of human IL-18.

Brand:  Invitrogen™ BMS267-2TEN

 View more versions of this product

Product Code. 15521857


Includes:
  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-18
  • Biotin-Conjugate anti-human IL-18 monoclonal antibody
  • Streptavidin-HRP
  • Human IL-18 Standard lyophilized, 10ng/mL upon reconstitution
  • Control high, lyophilized
  • Control low, lyophilized
  • Sample Diluent
  • Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
  • Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Blue-Dye
  • Green-Dye
  • Red-Dye
  • Adhesive Films
This item is not returnable. View return policy

Description

Description

The Human Interleukin-18 (Hu IL-18) ELISA quantitates Hu IL-18 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-18. Principle of the method The Human IL-18 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q14116
9.0 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
IGIF,IL-18,IL-1g,IL1F4
6.5%
HRP
RUO
2°C to 8°C
Human
3 hr. 10 min.
78-5,000 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
3606
8.1%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Interleukin-18, Interferon-gamma-inducing factor
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only