Learn More
Human IL-2 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-2 capture antibody and a biotinylated anti-IL-2 detection antibody
Brand: BD Biosciences 551884
Additional Details : Weight : 0.02000kg
Includes: Human IL-2 ELISPOT Capture Antibody (no azide/low endotoxin), Human IL-2 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Specifications
5 x 96-well Plates |