missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-35 Recombinant Protein, PeproTech®
HEK293 cells Recombinant Protein
£65.00 - £4170.00
Specifications
Accession Number | P29459 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3592 |
Molecular Weight (g/mol) | 45.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17881203
|
Gibco™
200-37-100UG |
2 x 50 μg | N/A | ||||||
17880163
|
Gibco™
200-37-10UG |
10 μg |
£158.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17821223
|
Gibco™
200-37-1MG |
20 x 50 μg | N/A | ||||||
17831313
|
Gibco™
200-37-250UG |
5 x 50 μg | N/A | ||||||
17870353
|
Gibco™
200-37-2UG |
2 μg |
£65.00
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850233
|
Gibco™
200-37-500UG |
10 x 50 μg | N/A | ||||||
17820743
|
Gibco™
200-37-50UG |
50 μg |
£506.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-35, or Interleukin 35 is a cytokine belonging to the IL12 family, that is produced by regulatory T cells. It is a dimeric protein composed of IL-12-alpha and IL-27-beta chains. Members of this class of cytokines are important in influencing T helper cell differentiation and supressing inflammatory response. IL-35 is known to induce proliferation of regulatory T cells, and reduce the activity of helper Th17 cells.Specifications
P29459 | |
protein with no preservative | |
45.8 | |
HEK293 cells | |
RUO | |
CLMF; CLMF p35; cytokine receptor; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; EBI3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; epstein-Barr virus-induced gene 3 protein; IL-12 subunit p35; IL-12, subunit p35; IL12A; IL-12 A; IL12p35; IL27 subunit; IL-27 subunit beta; IL27B; IL-27 B; il35; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12 A; interleukin 12 A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; interleukin-12 subunit alpha; Interleukin-27 subunit beta; Interleukin-35; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
3592 | |
Human IL-35 | |
-20°C | |
<1 EU/μg | |
IL12A | |
Unconjugated | |
Alpha Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS Beta Subunit: RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title