missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-4, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£177.00 - £2795.00
Specifications
Accession Number | P05112 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3565 |
Molecular Weight (g/mol) | 15.1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17853643
|
Gibco™
AF-200-04-100UG |
100 μg |
£582.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17853213
|
Gibco™
AF-200-04-1MG |
1 mg |
£2795.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17833003
|
Gibco™
AF-200-04-20UG |
20 μg |
£177.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17873403
|
Gibco™
AF-200-04-500UG |
500 μg |
£1674.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
P05112 | |
protein with no preservative | |
15.1 | |
E. coli | |
RUO | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4 delta2; interleukin 4 variant IL-4 delta3; interleukin 4 variant IL-4 int2A; interleukin 4 variant IL-4 int3A; interleukin 4 variant IL-4 int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
3565 | |
Human IL-4, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
Il4 | |
Unconjugated | |
MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |