Learn More
Invitrogen™ IL-5 Human ELISA Kit
For the quantitative detection of human IL-5.
Brand: Invitrogen™ BMS278TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes:
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-5
- Biotin-Conjugate anti-human IL-5 monoclonal antibody
- Streptavidin-HRP
- Human IL-5 Standard lyophilized, 1ng/mL upon reconstitution
- Sample Diluent
- Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (1M Phosphoric acid)
- Blue-Dye
- Green-Dye
- Red-Dye
- Adhesive Films
Description
The Human Interleukin-5 (Hu IL-5) ELISA quantitates Hu IL-5 in human serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-5. Principle of the method The Human IL-5 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.Specifications
P05113 | |
1.5 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
EDF,IL-5,TRF | |
6.6% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
7.8-500 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
3567 | |
6.8% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL; Supernatant, 50 μL | |
B cell differentiation factor 1, TRF, EDF | |
1 hr. 20 min. |
For Research Use Only